Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284400.2 | NP_001271329.1 | 9 | 3596 | ||
NM_001321675.1 | NP_001308604.1 | 10 | 3752 | |||
NM_001321676.1 | NP_001308605.1 | 11 | 3744 | |||
NM_001321677.1 | NP_001308606.1 | 9 | 3549 | |||
NM_001321678.1 | NP_001308607.1 | 9 | 3444 | |||
NM_001321679.1 | NP_001308608.1 | 9 | 3715 | |||
NM_001321680.1 | NP_001308609.1 | 9 | 3711 | |||
NM_144598.4 | NP_653199.2 | 10 | 3756 | |||
NR_135753.1 | 10 | 3661 | ||||
NR_135754.1 | 10 | 3509 | ||||
NR_135755.1 | 9 | 3616 | ||||
NR_135756.1 | 9 | 3620 | ||||
NR_135757.1 | 11 | 3730 | ||||
NR_135758.1 | 10 | 3784 | ||||
NR_135759.1 | 8 | 3504 |