Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284400.2 | NP_001271329.1 | 9 | 3881 | ||
NM_001321675.1 | NP_001308604.1 | 10 | 4037 | |||
NM_001321676.1 | NP_001308605.1 | 11 | 4029 | |||
NM_001321677.1 | NP_001308606.1 | 9 | 3834 | |||
NM_001321678.1 | NP_001308607.1 | 9 | 3729 | |||
NM_001321679.1 | NP_001308608.1 | 9 | 4000 | |||
NM_001321680.1 | NP_001308609.1 | 9 | 3996 | |||
NM_144598.4 | NP_653199.2 | 10 | 4041 | |||
NR_135753.1 | 10 | 3946 | ||||
NR_135754.1 | 10 | 3794 | ||||
NR_135755.1 | 9 | 3901 | ||||
NR_135756.1 | 9 | 3905 | ||||
NR_135757.1 | 11 | 4015 | ||||
NR_135758.1 | 10 | 4069 | ||||
NR_135759.1 | 8 | 3789 |