Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284400.2 | NP_001271329.1 | 9 | 3295 | ||
NM_001321675.1 | NP_001308604.1 | 10 | 3451 | |||
NM_001321676.1 | NP_001308605.1 | 11 | 3443 | |||
NM_001321677.1 | NP_001308606.1 | 9 | 3248 | |||
NM_001321678.1 | NP_001308607.1 | 9 | 3143 | |||
NM_001321679.1 | NP_001308608.1 | 9 | 3414 | |||
NM_001321680.1 | NP_001308609.1 | 9 | 3410 | |||
NM_144598.4 | NP_653199.2 | 10 | 3455 | |||
NR_135753.1 | 10 | 3360 | ||||
NR_135754.1 | 10 | 3208 | ||||
NR_135755.1 | 9 | 3315 | ||||
NR_135756.1 | 9 | 3319 | ||||
NR_135757.1 | 11 | 3429 | ||||
NR_135758.1 | 10 | 3483 | ||||
NR_135759.1 | 8 | 3203 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR593703.1 | - | 1 | 603 | Chr. 15 - 99387611 - 99389069 [+] on Build GRCh38 |