Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM208B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005252474.3 | XP_005252531.1 | 1 | 4728 | ||
XM_011519515.2 | XP_011517817.1 | 1 | 4728 | |||
XM_011519516.2 | XP_011517818.1 | 1 | 4728 | |||
XM_011519519.2 | XP_011517821.1 | 1 | 4728 | |||
XM_011519520.2 | XP_011517822.1 | 1 | 4728 | |||
XM_017016361.1 | XP_016871850.1 | 1 | 4728 | |||
GenBank mRNA | AL049233.1 | - | 1 | 2836 |