Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321265.1 | NP_001308194.1 | 12 | 6317 | ||
NM_001321266.1 | NP_001308195.1 | 11 | 6239 | |||
NM_001321267.1 | NP_001308196.1 | 14 | 6540 | |||
NM_138799.3 | NP_620154.2 | 13 | 6463 | |||
NR_135598.1 | 14 | 6515 | ||||
NR_135599.1 | 13 | 6782 | ||||
NR_135600.1 | 13 | 6369 | ||||
NR_135601.1 | 11 | 6158 | ||||
NR_135602.1 | 12 | 6330 | ||||
NR_135603.1 | 14 | 6556 | ||||
NR_135604.1 | 13 | 6479 | ||||
NR_135605.1 | 12 | 6291 | ||||
NR_135606.1 | 13 | 6382 | ||||
GenBank mRNA | AF052107.1 | - | 1 | 332 |