Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011536389.2 | XP_011534691.1 | 13 | 3991 | ||
GenBank mRNA | AK054781.1 | - | 1 | 1947 | ||
AK055388.1 | - | 12 | 2994 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA223442.1 | - | 1 | 661 | Chr. 14 - 50979074 - 50979754 [-] on Build GRCh38 |