Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324512.1 | NP_001311441.1 | 22 | 7595 | ||
NM_030955.3 | NP_112217.2 | 24 | 7850 | |||
XM_017009905.1 | XP_016865394.1 | 25 | 7949 | |||
XM_017009906.1 | XP_016865395.1 | 24 | 7350 | |||
XM_017009907.1 | XP_016865396.1 | 17 | 6242 | |||
XM_017009908.1 | XP_016865397.1 | 14 | 5834 | |||
XM_017009909.1 | XP_016865398.1 | 13 | 5748 | |||
GenBank mRNA | AK001058.1 | - | 1 | 708 |