Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321868.1 | NP_001308797.1 | 26 | 9650 | ||
NM_001321877.1 | NP_001308806.1 | 25 | 9580 | |||
NM_001321897.1 | NP_001308826.1 | 26 | 9477 | |||
NM_178423.2 | NP_848510.1 | 26 | 9534 | |||
NM_178425.3 | NP_848512.1 | 25 | 9398 | |||
XM_011515629.2 | XP_011513931.1 | 26 | 9492 | |||
XM_011515631.2 | XP_011513933.1 | 27 | 9806 | |||
XM_011515634.2 | XP_011513936.1 | 25 | 9360 | |||
XM_011515635.2 | XP_011513937.1 | 27 | 10358 | |||
XM_011515636.2 | XP_011513938.1 | 26 | 10079 | |||
XM_011515637.2 | XP_011513939.1 | 26 | 9503 | |||
XM_011515638.2 | XP_011513940.1 | 26 | 10073 | |||
XM_011515639.2 | XP_011513941.1 | 26 | 10270 | |||
XM_011515641.2 | XP_011513943.1 | 27 | 10355 | |||
XM_011515642.2 | XP_011513944.1 | 25 | 11084 | |||
XM_011515644.2 | XP_011513946.1 | 27 | 10330 | |||
XM_011515645.2 | XP_011513947.1 | 25 | 11069 | |||
XM_011515646.2 | XP_011513948.1 | 25 | 11060 | |||
XM_011515648.2 | XP_011513950.1 | 26 | 10088 | |||
XM_011515649.2 | XP_011513951.1 | 26 | 10292 | |||
XM_017012826.1 | XP_016868315.1 | 26 | 10064 | |||
XM_017012827.1 | XP_016868316.1 | 27 | 10147 | |||
XM_017012828.1 | XP_016868317.1 | 25 | 10140 |