Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321868.1 | NP_001308797.1 | 26 | 9106 | ||
NM_001321877.1 | NP_001308806.1 | 25 | 9036 | |||
NM_001321897.1 | NP_001308826.1 | 26 | 8933 | |||
NM_178423.2 | NP_848510.1 | 26 | 8990 | |||
NM_178425.3 | NP_848512.1 | 25 | 8854 | |||
XM_011515629.2 | XP_011513931.1 | 26 | 8948 | |||
XM_011515631.2 | XP_011513933.1 | 27 | 9262 | |||
XM_011515634.2 | XP_011513936.1 | 25 | 8816 | |||
XM_011515635.2 | XP_011513937.1 | 27 | 9814 | |||
XM_011515636.2 | XP_011513938.1 | 26 | 9535 | |||
XM_011515637.2 | XP_011513939.1 | 26 | 8959 | |||
XM_011515638.2 | XP_011513940.1 | 26 | 9529 | |||
XM_011515639.2 | XP_011513941.1 | 26 | 9726 | |||
XM_011515641.2 | XP_011513943.1 | 27 | 9811 | |||
XM_011515642.2 | XP_011513944.1 | 25 | 10540 | |||
XM_011515644.2 | XP_011513946.1 | 27 | 9786 | |||
XM_011515645.2 | XP_011513947.1 | 25 | 10525 | |||
XM_011515646.2 | XP_011513948.1 | 25 | 10516 | |||
XM_011515648.2 | XP_011513950.1 | 26 | 9544 | |||
XM_011515649.2 | XP_011513951.1 | 26 | 9748 | |||
XM_017012826.1 | XP_016868315.1 | 26 | 9520 | |||
XM_017012827.1 | XP_016868316.1 | 27 | 9603 | |||
XM_017012828.1 | XP_016868317.1 | 25 | 9596 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR622072.1 | - | 1 | 822 | Chr. 7 - 19000844 - 19002405 [+] on Build GRCh38 |