Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135253.1 | 2 | 1292 | |||
GenBank mRNA | BC042084.1 | - | 2 | 1292 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000555299.1 | - | 2 | 1292 | Chr. 14 - 92905697 - 92907482 [-] on Build GRCh38 |