Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 26237 | ||
NM_001164343.2 | NP_001157815.1 | 12 | 26699 | |||
NM_001164344.2 | NP_001157816.1 | 8 | 26663 | |||
NM_001164345.2 | NP_001157817.1 | 7 | 26617 | |||
NM_001164346.2 | NP_001157818.1 | 6 | 26256 | |||
NM_001164347.2 | NP_001157819.1 | 6 | 26278 | |||
NM_015642.5 | NP_056457.3 | 10 | 26461 | |||
NR_121662.1 | 7 | 24719 | ||||
GenBank mRNA | AL049423.1 | - | 1 | 1015 | ||
BC035518.1 | - | 1 | 1474 |