Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040651.1 | - | 1 | 1182 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM932275.1 | - | 1 | 36 | Chr. 10 - 1019331 - 1019938 [+] on Build GRCh38 |