Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286238.1 | NP_001273167.1 | 13 | 3800 | ||
NM_001286239.1 | NP_001273168.1 | 12 | 3806 | |||
NM_019073.3 | NP_061946.1 | 13 | 3848 | |||
XM_006710700.2 | XP_006710763.1 | 11 | 3647 | |||
XM_011541606.1 | XP_011539908.1 | 11 | 3557 | |||
XM_011541607.2 | XP_011539909.1 | 10 | 3411 | |||
XM_011541608.1 | XP_011539910.1 | 12 | 3785 | |||
XM_011541609.2 | XP_011539911.1 | 11 | 3493 | |||
XM_017001510.1 | XP_016856999.1 | 10 | 3597 | |||
XM_017001511.1 | XP_016857000.1 | 12 | 3750 | |||
XM_017001512.1 | XP_016857001.1 | 14 | 3691 | |||
XM_017001513.1 | XP_016857002.1 | 11 | 3455 | |||
XM_017001514.1 | XP_016857003.1 | 14 | 3733 | |||
XM_017001515.1 | XP_016857004.1 | 12 | 3578 | |||
GenBank mRNA | BC040655.1 | - | 1 | 259 |