Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525901.1 | XP_011524203.1 | 2 | 877 | ||
GenBank mRNA | AK097938.1 | - | 1 | 1261 |