Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304952.1 | NP_001291881.1 | 1, 2 | 218 | ||
GenBank mRNA | AK301868.1 | - | 1, 2 | 215 | ||
AY601916.1 | - | 1, 2 | 242 | |||
DB038438.1 | - | 1, 2 | 242 |