Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535471.1 | XP_011533773.1 | 3 | 1949 | ||
GenBank mRNA | AK023829.1 | - | 1 | 5124 |