Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC283454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094730.1 | - | 1 | 492 |
Gene Symbol: HRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020146.1 | XP_016875635.1 | 5 | 4550 |