Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002563.4 | NP_002554.1 | 1 | 5240 | ||
GenBank mRNA | AK026659.1 | - | 1 | 543 |
Set Membership: |
G-protein coupled receptor |