Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535040.2 | XP_011533342.1 | 19 | 3465 | ||
GenBank mRNA | AF086440.1 | - | 1 | 51 |