Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005465.4 | NP_005456.1 | 13 | 6620 | ||
XM_005272994.4 | XP_005273051.1 | 14 | 6845 | |||
XM_005272995.2 | XP_005273052.1 | 14 | 6651 | |||
GenBank mRNA | U79271.1 | - | 1 | 723 |