Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322177.1 | NP_001309106.1 | 2 | 4834 | ||
NM_177966.6 | NP_808881.3 | 3 | 4661 | |||
GenBank mRNA | U79276.1 | - | 1 | 796 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | EB386946.1 | - | 1 | 505 | Chr. 3 - 57562198 - 57563037 [+] on Build GRCh38 |