Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111907.1 | 2 | 1930 | |||
GenBank mRNA | AK126649.1 | - | 1 | 947 |