Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008206.1 | XP_016863695.1 | 10 | 2942 | ||
XM_017008207.1 | XP_016863696.1 | 9 | 2912 | |||
XM_017008208.1 | XP_016863697.1 | 12 | 3455 | |||
GenBank mRNA | AK129914.1 | - | 1 | 628 |