Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534875.2 | XP_011533177.1 | 3 | 2781 | ||
XM_011534878.2 | XP_011533180.1 | 3 | 2943 | |||
XM_011534879.2 | XP_011533181.1 | 3 | 2908 | |||
GenBank mRNA | AK000205.1 | - | 1 | 66 |