Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007838.1 | XP_016863327.1 | 7 | 1757 | ||
GenBank mRNA | AK002032.1 | - | 1 | 1936 |