Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1AIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199260.1 | NP_001186189.1 | 7 | 5764 | ||
NM_145034.4 | NP_659471.1 | 6 | 5345 | |||
XM_005244939.4 | XP_005244996.1 | 7 | 5737 | |||
XM_017000474.1 | XP_016855963.1 | 8 | 5882 | |||
XM_017000475.1 | XP_016855964.1 | 6 | 5606 | |||
XM_017000476.1 | XP_016855965.1 | 3 | 5046 | |||
GenBank mRNA | AK002036.1 | - | 1 | 667 |