Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271946.1 | NP_001258875.1 | 36 | 9193 | ||
NM_003062.3 | NP_003053.1 | 36 | 9172 | |||
XM_017009779.1 | XP_016865268.1 | 36 | 8575 | |||
GenBank mRNA | BC062365.1 | - | 1 | 908 |