Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038271.1 | 1 | 412 | |||
NR_038272.1 | 2 | 185 | ||||
GenBank mRNA | AK093733.1 | - | 1 | 379 | ||
BC041357.1 | - | 2 | 193 | |||
BC070113.1 | - | 2 | 188 | |||
DB229458.1 | - | 1 | 412 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB108507.1 | - | 1 | 379 | Chr. 2 - 178523224 - 178597615 [+] on Build GRCh38 |