Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038271.1 | 4, 5 | 793 | |||
GenBank mRNA | AK093733.1 | - | 4, 5 | 760 | ||
AK094031.1 | - | 1 | 324 |