Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533806.2 | XP_011532108.1 | 11 | 8999 | ||
XM_011533807.1 | XP_011532109.1 | 11 | 8884 | |||
XM_011533808.1 | XP_011532110.1 | 11 | 8670 | |||
XM_011533809.2 | XP_011532111.1 | 12 | 8596 | |||
XM_011533810.2 | XP_011532112.1 | 12 | 6639 | |||
XM_011533812.2 | XP_011532114.1 | 9 | 8843 | |||
XM_011533813.2 | XP_011532115.1 | 11 | 9037 | |||
XM_011533814.2 | XP_011532116.1 | 11 | 8961 | |||
GenBank mRNA | AF074992.1 | - | 1 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX113747.1 | - | 1 | 497 | Chr. 3 - 50588537 - 50589050 [+] on Build GRCh38 |