Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318714.1 | NP_001305643.1 | 4 | 865 | ||
GenBank mRNA | AK092013.1 | - | 1 | 580 | ||
BC034234.1 | - | 4 | 805 | 5296397 |