Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023225.1 | XP_016878714.1 | 1 | 363 | ||
GenBank mRNA | AK092013.1 | - | 1 | 985 |