Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FENDRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036444.1 | 4 | 923 | |||
GenBank mRNA | AK097312.1 | - | 1 | 834 | ||
AL833740.1 | - | 1 | 506 | |||
BX538111.1 | - | 2 | 1123 |