Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321635.1 | NP_001308564.1 | 11 | 3620 | ||
NM_001321636.1 | NP_001308565.1 | 10 | 3525 | |||
NM_024759.2 | NP_079035.1 | 12 | 3648 | |||
NR_135745.1 | 12 | 3784 | ||||
NR_135746.1 | 13 | 3906 | ||||
NR_135747.1 | 12 | 3844 | ||||
GenBank mRNA | AK025015.1 | - | 1 | 1418 |