Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023016.1 | XP_016878505.1 | 8 | 7574 | ||
XM_017023017.1 | XP_016878506.1 | 8 | 7473 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK126695.1 | - | 1 | 1579 | Chr. 16 - 89232866 - 89237071 [+] on Build GRCh38 |