Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011520238.2 | XP_011518540.1 | 8 | 9165 | ||
XM_011520239.2 | XP_011518541.1 | 9 | 9231 | |||
XM_011520240.2 | XP_011518542.1 | 7 | 9085 | |||
XM_011520241.2 | XP_011518543.1 | 4 | 9800 | |||
XM_011520242.2 | XP_011518544.1 | 4 | 9786 | |||
XM_011520243.2 | XP_011518545.1 | 7 | 9112 | |||
XM_011520244.2 | XP_011518546.1 | 6 | 9032 | |||
XM_017018070.1 | XP_016873559.1 | 8 | 9161 | |||
XM_017018071.1 | XP_016873560.1 | 8 | 9151 | |||
XM_017018072.1 | XP_016873561.1 | 8 | 9014 | |||
XM_017018073.1 | XP_016873562.1 | 6 | 8873 | |||
XM_017018074.1 | XP_016873563.1 | 4 | 9781 | |||
XM_017018075.1 | XP_016873564.1 | 5 | 8820 | |||
XM_017018076.1 | XP_016873565.1 | 3 | 9747 | |||
XM_017018077.1 | XP_016873566.1 | 6 | 8900 | |||
XM_017018078.1 | XP_016873567.1 | 5 | 9866 | |||
XM_017018079.1 | XP_016873568.1 | 6 | 9028 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR610091.1 | - | 1 | 1265 | Chr. 11 - 40107244 - 40109881 [-] on Build GRCh38 |