Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_137195.1 | 1 | 227 | |||
NR_137196.1 | 1 | 227 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK095670.1 | - | 1 | 120 | Chr. 11 - 122103422 - 122105509 [-] on Build GRCh38 |