Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017616.1 | XP_016873105.1 | 8 | 3534 | ||
XM_017017619.1 | XP_016873108.1 | 7 | 3388 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK055836.1 | - | 1 | 1140 | Chr. 11 - 93763063 - 93764659 [+] on Build GRCh38 |