Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023349.1 | XP_016878838.1 | 9 | 1407 | ||
XM_017023351.1 | XP_016878840.1 | 10 | 1486 | |||
XM_017023353.1 | XP_016878842.1 | 9 | 1539 | |||
XM_017023354.1 | XP_016878843.1 | 8 | 1255 | |||
XM_017023356.1 | XP_016878845.1 | 9 | 1443 | |||
GenBank mRNA | AK123247.1 | - | 1 | 925 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM719917.1 | - | 1 | 446 | Chr. 16 - 15699638 - 15700159 [+] on Build GRCh38 |