Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201480.1 | NP_001188409.1 | 26 | 4772 | ||
NM_001201481.1 | NP_001188410.1 | 24 | 4604 | |||
NM_001201482.1 | NP_001188411.1 | 24 | 4589 | |||
NM_032523.3 | NP_115912.1 | 25 | 4697 | |||
NM_145739.2 | NP_665682.1 | 24 | 4285 | |||
XM_011510539.1 | XP_011508841.1 | 25 | 4675 | |||
XM_017003265.1 | XP_016858754.1 | 27 | 4844 | |||
XM_017003266.1 | XP_016858755.1 | 27 | 4844 | |||
XM_017003267.1 | XP_016858756.1 | 25 | 4539 | |||
XM_017003268.1 | XP_016858757.1 | 25 | 4660 | |||
XM_017003269.1 | XP_016858758.1 | 24 | 4431 | |||
XM_017003270.1 | XP_016858759.1 | 25 | 4758 | |||
XM_017003271.1 | XP_016858760.1 | 23 | 4356 | |||
XM_017003272.1 | XP_016858761.1 | 24 | 4567 | |||
XM_017003273.1 | XP_016858762.1 | 23 | 4507 | |||
XM_017003274.1 | XP_016858763.1 | 23 | 4492 | |||
XM_017003275.1 | XP_016858764.1 | 24 | 4586 | |||
XM_017003276.1 | XP_016858765.1 | 23 | 4492 | |||
XM_017003277.1 | XP_016858766.1 | 22 | 4399 | |||
GenBank mRNA | AK123248.1 | - | 1 | 379 |