Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015983.3 | NP_057067.1 | 7 | 3325 | ||
XM_006715734.3 | XP_006715797.1 | 8 | 2912 | |||
XM_011515420.2 | XP_011513722.1 | 8 | 3418 | |||
XM_011515422.2 | XP_011513724.1 | 6 | 3262 | |||
XM_011515423.2 | XP_011513725.1 | 5 | 2696 | |||
XM_017012292.1 | XP_016867781.1 | 9 | 3005 | |||
XM_017012295.1 | XP_016867784.1 | 6 | 2759 | |||
GenBank mRNA | AK056566.1 | - | 1 | 1942 |