Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011308.1 | XP_016866797.1 | 9 | 2182 | ||
GenBank mRNA | AK123370.1 | - | 1 | 949 | ||
AK123429.1 | - | 1 | 2026 | |||
BC040707.1 | - | 1 | 995 |