Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297605.1 | NP_001284534.1 | 7 | 2811 | ||
NM_003820.3 | NP_003811.2 | 8 | 2954 | |||
GenBank mRNA | AK130172.1 | - | 1 | 1268 |