Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128796.1 | - | 1 | 1566 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA246593.1 | - | 1 | 348 | Chr. 12 - 53174639 - 53175210 [-] on Build GRCh38 |