Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011553.3 | NP_001011553.2 | 13 | 4204 | ||
NM_001242956.1 | NP_001229885.1 | 12 | 4099 | |||
NM_001788.5 | NP_001779.3 | 14 | 4207 |