Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542594.2 | XP_011540896.1 | 8 | 3860 | ||
XM_011542595.2 | XP_011540897.1 | 9 | 3972 | |||
XM_011542596.2 | XP_011540898.1 | 9 | 4213 | |||
XM_011542597.2 | XP_011540899.1 | 8 | 4101 | |||
XM_011542598.2 | XP_011540900.1 | 7 | 3801 | |||
XM_011542599.2 | XP_011540901.1 | 7 | 4042 | |||
GenBank mRNA | AK127883.1 | - | 1 | 710 |