Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC162137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029861.1 | - | 1 | 1739 | 5167909 |
Gene Symbol: LOC107984902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023962.1 | XP_016879451.1 | 1 | 482 |
Gene Symbol: LOC107987236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023961.1 | XP_016879450.1 | 1 | 482 |