Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005252642.4 | XP_005252699.1 | 12 | 2531 | ||
XM_017016949.1 | XP_016872438.1 | 13 | 2470 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK025065.1 | - | 1 | 944 | Chr. 10 - 15104709 - 15106707 [-] on Build GRCh38 |