Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055653.1 | - | 45 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA603424.1 | - | 1 | 45 | Chr. 3 - 42622499 - 42623292 [+] on Build GRCh38 |