Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028444.1 | XP_016883933.1 | 7 | 2477 | ||
XM_017028446.1 | XP_016883935.1 | 6 | 2337 |